The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173097.1 | NP_775120.1 | 5 - 6 | 478 | 119 |
XM_008763019.1 | XP_008761241.1 | 5 - 6 | 616 | 119 | |
GenBank mRNA | L20869.1 | - | 5 - 6 | 478 | 119 |