Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Asgr1
Entrez Gene ID
24210
Gene Name
asialoglycoprotein receptor 1
Gene Aliases
ASGR, RATRHL1, RHL1
Chromosome Location
Chr.10: 56662191 - 56666086 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012503.2 NP_036635.1 5 - 6 417 90
XM_006246579.1 XP_006246641.1 5 - 6 529 90
XM_008767768.1 XP_008765990.1 5 - 6 549 90
XM_008767769.1 XP_008765991.1 5 - 6 493 90
GenBank mRNA BC088154.1 - 5 - 6 417 90
FQ209763.1 - 5 - 6 453 90
FQ218553.1 - 5 - 6 493 90
FQ218644.1 - 5 - 6 453 90
FQ218824.1 - 5 - 6 453 90
FQ219107.1 - 5 - 6 490 90
FQ219710.1 - 5 - 6 534 90
M21770.1 - 4 - 5 319 90

View Full Product Details