The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017038.1 | NP_058734.1 | 5 - 6 | 1208 | 103 |
XM_006240193.2 | XP_006240255.1 | 5 - 6 | 4511 | 103 | |
XM_006240194.2 | XP_006240256.1 | 5 - 6 | 1465 | 103 | |
XM_006240195.2 | XP_006240257.1 | 5 - 6 | 1229 | 103 | |
GenBank mRNA | BC097333.1 | - | - | 638 | 103 |
J04503.1 | - | 5 - 6 | 1208 | 103 |