Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfpi
Entrez Gene ID
29436
Gene Name
tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor)
Gene Aliases
-
Chromosome Location
Chr.3: 71852738 - 71902126 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017200.1 NP_058896.1 6 - 7 730 79
XM_006234440.2 XP_006234502.1 7 - 8 1025 79
XM_006234441.2 XP_006234503.1 7 - 8 2046 79
XM_006234444.2 XP_006234506.1 7 - 8 2059 79
XM_008761980.1 XP_008760202.1 7 - 8 1044 79
XM_008761981.1 XP_008760203.1 7 - 8 808 79
XM_008761982.1 XP_008760204.1 7 - 8 1038 79
XM_008761983.1 XP_008760205.1 7 - 8 2061 79
XM_008761984.1 XP_008760206.1 7 - 8 2074 79
GenBank mRNA D10926.1 - 6 - 7 730 79
FQ219876.1 - 4 - 5 436 79
FQ220339.1 - 4 - 5 430 79
FQ220655.1 - 4 - 5 527 79
FQ220711.1 - 4 - 5 527 79
FQ229364.1 - 4 - 5 459 79

View Full Product Details