The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017270.2 | NP_058966.2 | 8 - 9 | 1157 | 61 |
GenBank mRNA | BC127504.1 | - | 8 - 9 | 1157 | 61 |
X90710.1 | - | 8 - 9 | 1183 | 61 |