Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psma1
Entrez Gene ID
29668
Gene Name
proteasome (prosome, macropain) subunit, alpha type 1
Gene Aliases
-
Chromosome Location
Chr.1: 183733567 - 183744586 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017278.1 NP_058974.1 9 - 10 822 150
GenBank mRNA BC062233.1 - 9 - 10 819 150
D90265.1 - 9 - 10 822 150
FQ212620.1 - 9 - 10 791 150
FQ213606.1 - 9 - 10 813 150
FQ215179.1 - 9 - 10 801 150
FQ215378.1 - 9 - 10 805 150
FQ216107.1 - 9 - 10 815 150
FQ218243.1 - - 854 150
FQ220925.1 - 9 - 10 787 150
FQ225726.1 - 9 - 10 773 150
FQ229288.1 - 9 - 10 849 150
M29859.1 - 9 - 10 822 150

View Full Product Details