The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017294.1 | NP_058990.1 | 9 - 10 | 1374 | 65 |
GenBank mRNA | AF104402.1 | - | 9 - 10 | 1374 | 65 |