Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Calca
Entrez Gene ID
24241
Gene Name
calcitonin-related polypeptide alpha
Gene Aliases
CAL6, CGRP, Cal1, Calc, RATCAL6, calcitonin
Chromosome Location
Chr.1: 184184018 - 184188938 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017338.2 NP_059034.1 3 - 4 336 70
XM_008759676.1 XP_008757898.1 3 - 4 387 70
GenBank mRNA M26137.1 - 2 - 3 241 70
V01228.1 - 3 - 4 297 70
V01229.1 - 2 - 3 241 70

View Full Product Details