Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Isl2
Entrez Gene ID
57233
Gene Name
ISL LIM homeobox 2
Gene Aliases
-
Chromosome Location
Chr.8: 60117565 - 60122716 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020471.1 NP_065204.1 5 - 6 1047 57
GenBank mRNA L35571.1 - 5 - 6 1047 57

View Full Product Details