The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175843.4 | NP_787037.2 | 3 - 4 | 556 | 86 |
NM_181550.1 | NP_853528.1 | 3 - 4 | 522 | 86 | |
XM_006246213.2 | XP_006246275.1 | 3 - 4 | 578 | 86 | |
XM_006246214.2 | XP_006246276.1 | 3 - 4 | 578 | 86 | |
XM_006246215.2 | XP_006246277.1 | 3 - 4 | 578 | 86 | |
GenBank mRNA | AF439403.1 | - | 3 - 4 | 522 | 86 |
BC061575.1 | - | 3 - 4 | 560 | 86 | |
Y08355.1 | - | 3 - 4 | 585 | 86 |