Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il1rapl1
Entrez Gene ID
317553
Gene Name
interleukin 1 receptor accessory protein-like 1
Gene Aliases
Il1rapl
Chromosome Location
Chr.X: 55439412 - 56765893 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177935.2 NP_808796.1 3 - 4 552 98
GenBank mRNA AY216593.1 - 3 - 4 552 98

View Full Product Details