The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178094.3 | NP_835195.1 | 3 - 4 | 1559 | 61 |
GenBank mRNA | AJ440782.1 | - | 3 - 4 | 1559 | 61 |
AY160770.1 | - | 3 - 4 | 1805 | 61 |