Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem184c
Entrez Gene ID
291946
Gene Name
transmembrane protein 184C
Gene Aliases
Tmem34
Chromosome Location
Chr.19: 34041165 - 34057165 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178330.1 NP_847900.1 3 - 4 753 145
XM_006255422.2 XP_006255484.1 4 - 5 465 145
XM_008772475.1 XP_008770697.1 4 - 5 1364 145
GenBank mRNA AY228474.1 - 3 - 4 753 145
BC089112.1 - 3 - 4 763 145

View Full Product Details