Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lhx9
Entrez Gene ID
289048
Gene Name
LIM homeobox 9
Gene Aliases
-
Chromosome Location
Chr.13: 55854543 - 55879305 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181367.2 NP_852032.1 6 - 7 1034 75
XM_006249914.2 XP_006249976.1 4 - 5 1526 75
XM_006249915.2 XP_006249977.1 4 - 5 1528 75
XM_006249916.2 XP_006249978.1 7 - 8 1596 75
XM_006249917.1 XP_006249979.1 4 - 5 887 75
GenBank mRNA AF527619.1 - 4 - 5 842 75
AY273890.1 - 6 - 7 1034 75
AY724502.1 - 6 - 7 1034 75

View Full Product Details