Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nit1
Entrez Gene ID
289222
Gene Name
nitrilase 1
Gene Aliases
-
Chromosome Location
Chr.13: 89722370 - 89725806 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082580.1 NP_001076049.1 4 - 5 566 79
NM_182668.2 NP_872609.2 4 - 5 534 79
GenBank mRNA AY300752.1 - 3 - 4 397 79
BC087146.1 - 4 - 5 515 79
CO558072.1 - 4 - 5 572 79
FQ214471.1 - - 564 79
FQ227281.1 - - 563 79

View Full Product Details