The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019323.1 | NP_062196.1 | - | 587 | 72 |
GenBank mRNA | U67912.1 | - | - | 568 | 72 |
U72143.1 | - | - | 587 | 72 |