The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053532.1 | NP_445984.1 | 1 - 2 | 68 | 87 |
GenBank mRNA | AF285103.1 | - | 1 - 2 | 68 | 87 |
BC060551.1 | - | 1 - 2 | 78 | 87 | |
FQ224099.1 | - | 1 - 2 | 87 |