Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Amigo2
Entrez Gene ID
300186
Gene Name
adhesion molecule with Ig like domain 2
Gene Aliases
Ali1
Chromosome Location
Chr.7: 138704124 - 138707221 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006242290.1 XP_006242352.1 1 - 1 1507 61
GenBank mRNA AY237730.1 - 1 - 1 1446 61

View Full Product Details