Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc50
Entrez Gene ID
288022
Gene Name
coiled-coil domain containing 50
Gene Aliases
C3orf6h, c3orf6
Chromosome Location
Chr.11: 76742179 - 76804698 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182736.1 NP_877400.1 8 - 9 718 134
XM_008768795.1 XP_008767017.1 9 - 10 1483 134
GenBank mRNA AJ534984.1 - 8 - 9 718 134

View Full Product Details