Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ndufa5
Entrez Gene ID
25488
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 5
Gene Aliases
NADHUO
Chromosome Location
Chr.4: 51590413 - 51598771 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012985.2 NP_037117.1 1 - 2 170 56
GenBank mRNA BC059148.1 - 1 - 2 33 56
D86215.1 - 1 - 2 170 56
FQ212236.1 - 1 - 2 56
FQ228773.1 - 1 - 2 31 56

View Full Product Details