Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
St13
Entrez Gene ID
81800
Gene Name
suppression of tumorigenicity 13
Gene Aliases
-
Chromosome Location
Chr.7: 122585770 - 122635731 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031122.1 NP_112384.1 5 - 6 445 60
GenBank mRNA BC078804.1 - 5 - 6 452 60
FQ212434.1 - - 502 60
FQ228746.1 - 5 - 6 407 60
X82021.1 - 5 - 6 445 60

View Full Product Details