The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031237.1 | NP_112516.1 | 5 - 6 | 575 | 81 |
XM_006233333.1 | XP_006233395.1 | 6 - 7 | 567 | 81 | |
XM_006233334.1 | XP_006233396.1 | 6 - 7 | 714 | 81 | |
XM_006233335.1 | XP_006233397.1 | 6 - 7 | 521 | 81 | |
XM_006233336.2 | XP_006233398.1 | 6 - 7 | 567 | 81 | |
XM_006233337.1 | XP_006233399.1 | 6 - 7 | 714 | 81 | |
XM_006233338.1 | XP_006233400.1 | 6 - 7 | 552 | 81 | |
GenBank mRNA | AB006852.1 | - | 5 - 6 | 575 | 81 |
AY724500.1 | - | 6 - 7 | 522 | 81 | |
BC072696.1 | - | 6 - 7 | 498 | 81 | |
FQ215509.1 | - | 1 - 2 | 102 | 81 | |
FQ226278.1 | - | 6 - 7 | 551 | 81 | |
FQ233355.1 | - | 6 - 7 | 542 | 81 | |
U13175.1 | - | 5 - 6 | 429 | 81 |