Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc25a4
Entrez Gene ID
85333
Gene Name
solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 4
Gene Aliases
Ant1
Chromosome Location
Chr.16: 49266903 - 49270698 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053515.1 NP_445967.1 3 - 4 847 77
GenBank mRNA BC060533.1 - 3 - 4 851 77
D12770.1 - 3 - 4 814 77
FQ213027.1 - 3 - 4 844 77
FQ213612.1 - 3 - 4 873 77
FQ215448.1 - 3 - 4 846 77
FQ217254.1 - 3 - 4 843 77
FQ220525.1 - 3 - 4 842 77
FQ224814.1 - 3 - 4 844 77

View Full Product Details