The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080902.4 | NP_543178.2 | 2 - 3 | 174 | 70 |
GenBank mRNA | AY062253.3 | - | 2 - 3 | 233 | 70 |
BC059118.1 | - | 2 - 3 | 178 | 70 |