The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134415.2 | NP_599242.2 | 3 - 4 | 437 | 71 |
GenBank mRNA | BC098657.1 | - | 3 - 4 | 428 | 71 |
DQ354588.1 | - | 3 - 4 | 427 | 71 | |
FQ212851.1 | - | 3 - 4 | 449 | 71 | |
FQ214845.1 | - | 3 - 4 | 456 | 71 | |
FQ225571.1 | - | 3 - 4 | 450 | 71 | |
FQ226635.1 | - | 3 - 4 | 464 | 71 | |
FQ229542.1 | - | 3 - 4 | 455 | 71 | |
Y17325.1 | - | 3 - 4 | 414 | 69 |