Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ctsr
Entrez Gene ID
290975
Gene Name
cathepsin R
Gene Aliases
Cts6
Chromosome Location
Chr.17: 3885684 - 3894447 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175581.3 NP_783171.2 6 - 7 866 95
XM_008771434.1 XP_008769656.1 5 - 6 787 95
GenBank mRNA AF456459.1 - - 826 95
BC097484.1 - 6 - 7 866 95

View Full Product Details