Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp11b3
Entrez Gene ID
353498
Gene Name
cytochrome P450, family 11, subfamily b, polypeptide 3
Gene Aliases
CYPXIB3
Chromosome Location
Chr.7: 116155928 - 116161949 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181824.1 NP_861545.1 - 797 72
XM_008765572.1 XP_008763794.1 4 - 5 1038 72
GenBank mRNA U14907.1 - - 797 72

Gene Symbol
LOC100910462
Entrez Gene ID
100910462
Gene Name
cytochrome P450 11B1, mitochondrial-like
Gene Aliases
-
Chromosome Location
Chr.7: 116195976 - 116201999 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750363.3 XP_003750411.2 4 - 5 1040 72
XM_008765623.1 XP_008763845.1 4 - 5 1038 72

View Full Product Details