The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017122.1 | NP_058818.1 | 2 - 3 | 657 | 63 |
XM_006238920.1 | XP_006238982.1 | 3 - 4 | 602 | 63 | |
XM_006238921.2 | XP_006238983.1 | 3 - 4 | 559 | 63 | |
GenBank mRNA | AY442172.1 | - | 2 - 3 | 484 | 63 |
BC087632.1 | - | 2 - 3 | 512 | 63 | |
D12573.1 | - | 2 - 3 | 657 | 63 |