Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mpped2
Entrez Gene ID
362185
Gene Name
metallophosphoesterase domain containing 2
Gene Aliases
C11orf8, C11orf8h
Chromosome Location
Chr.3: 97720308 - 97899735 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198778.2 NP_942073.2 3 - 4 462 90
XM_006234680.2 XP_006234742.1 3 - 4 1837 90
XM_006234681.2 XP_006234743.1 3 - 4 539 90
XM_006234682.2 XP_006234744.1 3 - 4 626 90
XM_006234683.2 XP_006234745.1 3 - 4 541 90
XM_008762104.1 XP_008760326.1 3 - 4 1261 90
XM_008762105.1 XP_008760327.1 2 - 3 1521 90
GenBank mRNA BC161828.1 - 3 - 4 544 90
EV763409.1 - 3 - 4 566 90
EV768250.1 - 1 - 2 97 90

View Full Product Details