The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106488.2 | NP_001099958.2 | 1 - 2 | 151 | 63 |
GenBank mRNA | BC158687.1 | - | 1 - 2 | 102 | 63 |
DY311538.1 | - | - | 102 | 63 | |
EV779017.1 | - | - | 172 | 63 |