Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab5b
Entrez Gene ID
288779
Gene Name
RAB5B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.7: 3104530 - 3123377 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006240750.2 XP_006240812.1 1 - 2 85 79

View Full Product Details