The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031025.1 | NP_112287.1 | 7 - 8 | 1093 | 71 |
GenBank mRNA | BC107440.1 | - | 7 - 8 | 1093 | 71 |
D00092.1 | - | 4 - 5 | 597 | 71 | |
D10655.1 | - | 6 - 7 | 810 | 71 | |
M16075.1 | - | 4 - 5 | 597 | 71 |