Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vstm2a
Entrez Gene ID
689106
Gene Name
V-set and transmembrane domain containing 2A
Gene Aliases
-
Chromosome Location
Chr.14: 99529013 - 99558304 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770441.1 XP_008768663.1 4 - 5 907 64

View Full Product Details