Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ldb1
Entrez Gene ID
309447
Gene Name
LIM domain binding 1
Gene Aliases
-
Chromosome Location
Chr.1: 265785371 - 265798442 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107601.1 NP_001101071.1 2 - 3 661 67
XM_006231493.1 XP_006231555.1 2 - 3 493 67
XM_006231494.1 XP_006231556.1 2 - 3 493 67
GenBank mRNA BC166757.1 - 2 - 3 298 67

View Full Product Details