Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc16a4
Entrez Gene ID
295356
Gene Name
solute carrier family 16, member 4
Gene Aliases
-
Chromosome Location
Chr.2: 210045118 - 210161027 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013913.1 NP_001013935.1 7 - 8 1433 93
XM_008761384.1 XP_008759606.1 7 - 8 1604 93
XM_008761385.1 XP_008759607.1 7 - 8 1607 93
XM_008761386.1 XP_008759608.1 7 - 8 1602 93
XM_008761387.1 XP_008759609.1 7 - 8 1602 93
XM_008761388.1 XP_008759610.1 6 - 7 1089 93
XM_008761389.1 XP_008759611.1 6 - 7 1077 93
XM_008761390.1 XP_008759612.1 7 - 8 1597 93
GenBank mRNA BC085920.1 - 7 - 8 1433 93

View Full Product Details