Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnip4
Entrez Gene ID
259243
Gene Name
Kv channel interacting protein 4
Gene Aliases
Kchip4
Chromosome Location
Chr.14: 65549366 - 66747926 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181365.2 NP_852030.1 - 801 68
GenBank mRNA AF345444.1 - - 801 68
AF345445.1 - 3 - 4 371 68
AF453245.1 - 4 - 5 424 68

View Full Product Details