Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nova1
Entrez Gene ID
298992
Gene Name
neuro-oncological ventral antigen 1
Gene Aliases
Nova-1
Chromosome Location
Chr.6: 66960126 - 67085844 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006240069.2 XP_006240131.1 4 - 5 966 70
XM_006240070.2 XP_006240132.1 4 - 5 850 70
XM_006240072.2 XP_006240134.1 3 - 4 1673 70

View Full Product Details