Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adck3
Entrez Gene ID
360887
Gene Name
aarF domain containing kinase 3
Gene Aliases
Cabc1
Chromosome Location
Chr.13: 98451629 - 98480438 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013185.1 NP_001013203.1 6 - 7 893 58
XM_006250387.1 XP_006250449.1 6 - 7 976 58
GenBank mRNA BC091388.1 - 6 - 7 893 58

View Full Product Details