The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013189.1 | NP_001013207.1 | 4 - 5 | 655 | 73 |
XM_006254952.2 | XP_006255014.1 | 5 - 6 | 1019 | 73 | |
XM_006254953.2 | XP_006255015.1 | 5 - 6 | 1412 | 73 | |
XM_006254954.1 | XP_006255016.1 | 3 - 4 | 1133 | 73 | |
XM_006254955.2 | XP_006255017.1 | 4 - 5 | 964 | 73 | |
XM_006254956.2 | XP_006255018.1 | 4 - 5 | 1355 | 73 | |
XM_008772155.1 | XP_008770377.1 | 4 - 5 | 1187 | 73 | |
GenBank mRNA | BC079072.1 | - | 4 - 5 | 1185 | 73 |
BC088186.1 | - | 4 - 5 | 655 | 73 |