Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnni3
Entrez Gene ID
29248
Gene Name
troponin I type 3 (cardiac)
Gene Aliases
TnI, cTNI
Chromosome Location
Chr.1: 72882806 - 72886490 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017144.2 NP_058840.1 3 - 4 213 70
GenBank mRNA DQ062462.1 - 1 - 2 109 70
M57679.1 - 3 - 4 196 70
M92074.1 - 2 - 3 163 70
X58499.1 - 3 - 4 214 70

View Full Product Details