Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhbdd1
Entrez Gene ID
316557
Gene Name
rhomboid domain containing 1
Gene Aliases
RGD1306477, RRP4, rRHBDD1
Chromosome Location
Chr.9: 88110041 - 88229462 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767250.1 XP_008765472.1 1 - 2 825 99

View Full Product Details