Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wnt10a
Entrez Gene ID
316527
Gene Name
wingless-type MMTV integration site family, member 10A
Gene Aliases
-
Chromosome Location
Chr.9: 82053581 - 82066047 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108227.1 NP_001101697.1 3 - 4 1410 65

View Full Product Details