Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Erc2
Entrez Gene ID
259269
Gene Name
ELKS/RAB6-interacting/CAST family member 2
Gene Aliases
CAST, Cmbp
Chromosome Location
Chr.16: 3293657 - 4283592 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170787.2 NP_740768.1 8 - 9 1771 113
GenBank mRNA AF541925.1 - 8 - 9 1771 113
AY049038.1 - 8 - 9 1973 113

View Full Product Details