Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc1a3
Entrez Gene ID
29483
Gene Name
solute carrier family 1 (glial high affinity glutamate transporter), member 3
Gene Aliases
EAAT1, GLAST, GLAST-1, GluT-1
Chromosome Location
Chr.2: 57860881 - 57935363 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289941.1 NP_001276870.1 2 - 3 461 93
GenBank mRNA AF265360.1 - 2 - 3 300 93

View Full Product Details