Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo4
Entrez Gene ID
310363
Gene Name
F-box protein 4
Gene Aliases
-
Chromosome Location
Chr.2: 53797954 - 53811134 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107672.1 NP_001101142.1 2 - 3 887 76

View Full Product Details