Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl2
Entrez Gene ID
290692
Gene Name
kelch-like family member 2
Gene Aliases
-
Chromosome Location
Chr.16: 26755241 - 26852134 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771199.1 XP_008769421.1 8 - 9 1190 65

View Full Product Details