Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csgalnact1
Entrez Gene ID
306375
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 1
Gene Aliases
Chgn, RGD1307618
Chromosome Location
Chr.16: 22704412 - 23075071 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107309.1 NP_001100779.1 2 - 3 842 136
XM_006253026.2 XP_006253088.1 3 - 4 1298 136
XM_006253027.2 XP_006253089.1 4 - 5 1393 136
XM_006253028.2 XP_006253090.1 4 - 5 1807 136
XM_006253029.2 XP_006253091.1 4 - 5 1613 136
XM_006253030.2 XP_006253092.1 4 - 5 1697 136
XM_006253031.2 XP_006253093.1 3 - 4 1643 136
XM_008771110.1 XP_008769332.1 4 - 5 1427 136
XM_008771111.1 XP_008769333.1 4 - 5 1432 136
XM_008771112.1 XP_008769334.1 4 - 5 1433 136
XM_008771113.1 XP_008769335.1 4 - 5 1432 136
XM_008771114.1 XP_008769336.1 4 - 5 1431 136
XM_008771115.1 XP_008769337.1 6 - 7 1567 136
XM_008771116.1 XP_008769338.1 4 - 5 1571 136
GenBank mRNA BC168857.1 - 2 - 3 1066 136

View Full Product Details