The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107309.1 | NP_001100779.1 | 6 - 7 | 1302 | 64 |
XM_006253026.2 | XP_006253088.1 | 7 - 8 | 1758 | 64 | |
XM_006253027.2 | XP_006253089.1 | 8 - 9 | 1853 | 64 | |
XM_006253028.2 | XP_006253090.1 | 8 - 9 | 2267 | 64 | |
XM_006253029.2 | XP_006253091.1 | 8 - 9 | 2073 | 64 | |
XM_006253030.2 | XP_006253092.1 | 8 - 9 | 2157 | 64 | |
XM_006253031.2 | XP_006253093.1 | 7 - 8 | 2103 | 64 | |
XM_008771110.1 | XP_008769332.1 | 8 - 9 | 1887 | 64 | |
XM_008771111.1 | XP_008769333.1 | 8 - 9 | 1892 | 64 | |
XM_008771112.1 | XP_008769334.1 | 8 - 9 | 1893 | 64 | |
XM_008771113.1 | XP_008769335.1 | 8 - 9 | 1892 | 64 | |
XM_008771114.1 | XP_008769336.1 | 8 - 9 | 1891 | 64 | |
XM_008771115.1 | XP_008769337.1 | 10 - 11 | 2027 | 64 | |
XM_008771116.1 | XP_008769338.1 | 8 - 9 | 2031 | 64 | |
GenBank mRNA | BC168857.1 | - | 6 - 7 | 1526 | 64 |