Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc22a14
Entrez Gene ID
316061
Gene Name
solute carrier family 22, member 14
Gene Aliases
-
Chromosome Location
Chr.8: 127858425 - 127871192 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108193.1 NP_001101663.1 2 - 3 619 64
GenBank mRNA BC168201.1 - - 722 64

View Full Product Details