Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cxadrl1
Entrez Gene ID
361873
Gene Name
coxsackie virus and adenovirus receptor-like 1
Gene Aliases
-
Chromosome Location
Chr.8: 107246656 - 107272996 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199406.2 NP_955438.1 6 - 7 1043 102
XM_008766539.1 XP_008764761.1 6 - 7 1830 102
XM_008766540.1 XP_008764762.1 5 - 6 1150 102
GenBank mRNA BC063181.1 - 6 - 7 1048 102

Gene Symbol
LOC103690070
Entrez Gene ID
103690070
Gene Name
coxsackievirus and adenovirus receptor-like
Gene Aliases
-
Chromosome Location
Chr.8: 106832986 - 106859328 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008766579.1 XP_008764801.1 - 1830 102
XM_008766580.1 XP_008764802.1 - 1830 102
XM_008766581.1 XP_008764803.1 - 1150 102

View Full Product Details